Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer powder pillows

Buffer powder pillows

Supplier:   Hach
Buffer powder pillows
Supplier:  Hach
Certificates
Buffer powder pillows
Buffer powder pillows

13 Options Available Below

Product Details & Documents

The unit-dose powder pillows assure buffer freshness and eliminate the possibility of contamination. Each powder pillow prepares 50 ml of buffer solutions of respective pH.

  • Fresh and reliable
  • No danger of contamination
  • Traceable to NIST standards

Delivery information: The buffer pillows are delivered in an airtight container and are supplied with a certificate of conformity.

Product Family Options

Product Information

  • pH value
    pH accuracy
    pH buffer type
    Colour
    Reference temperature
    Pk
    Availability
    Price
  • 6,86
    ±0,02
    Phosphate
    Colourless
    25 °C
    15
    Log in for availability
    Log in for availability
    1 * 15 ST
    1 * 15 ST - 25,50 €
    25,50 €
    1 * 15 ST
       25,50 €

  • 8,00
    ±0,02
    Phosphate
    Colourless
    25 °C
    15
    Log in for availability
    Log in for availability
    1 * 15 ST
    1 * 15 ST - 25,50 €
    25,50 €
    1 * 15 ST
       25,50 €

  • 9,00
    ±0,02
    Borate
    Colourless
    25 °C
    50
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 77,10 €
    77,10 €
    1 * 50 ST
       77,10 €

  • 4,01 and 7,00
    ±0,02
    Red/yellow
    25 °C
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 27,20 €
    27,20 €
    1 * 20 ST
       27,20 €

  • 4,01
    ±0,02
    Phthalate
    Red
    25 °C
    250
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 286,00 €
    286,00 €
    1 * 250 ST
       286,00 €

  • 4,01
    ±0,02
    Phthalate
    Red
    25 °C
    50
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 77,10 €
    77,10 €
    1 * 50 ST
       77,10 €

  • 4,01
    ±0,02
    Phthalate
    Red
    25 °C
    15
    Log in for availability
    Log in for availability
    1 * 15 ST
    1 * 15 ST - 25,50 €
    25,50 €
    1 * 15 ST
       25,50 €

  • 7,00
    ±0,02
    Phosphate
    Yellow
    25 °C
    250
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 286,00 €
    286,00 €
    1 * 250 ST
       286,00 €

  • 7,00
    ±0,02
    Phosphate
    Yellow
    25 °C
    50
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 77,10 €
    77,10 €
    1 * 50 ST
       77,10 €

  • 7,00
    ±0,02
    Phosphate
    Yellow
    25 °C
    15
    Log in for availability
    Log in for availability
    1 * 15 ST
    1 * 15 ST - 25,50 €
    25,50 €
    1 * 15 ST
       25,50 €

  • 10,01
    ±0,02
    Carbonate
    Blue
    25 °C
    250
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 286,00 €
    286,00 €
    1 * 250 ST
       286,00 €

  • 10,01
    ±0,02
    Carbonate
    Blue
    25 °C
    50
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 77,10 €
    77,10 €
    1 * 50 ST
       77,10 €

  • 10,01
    ±0,02
    Carbonate
    Blue
    25 °C
    15
    Log in for availability
    Log in for availability
    1 * 15 ST
    1 * 15 ST - 25,50 €
    25,50 €
    1 * 15 ST
       25,50 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.