ContactUsLinkComponent
Visual comparator test kits, nitrate-nitrite, NI-12
Visual comparator test kits, nitrate-nitrite, NI-12
Catalog # HACH1408100
Supplier:  Hach
CAS Number:  
undefined
Visual comparator test kits, nitrate-nitrite, NI-12
Catalog # HACH1408100
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Nitrate - Nitrite test kit, model NI-12
  • Type:
    Cadmium reduction, diazotisation
  • Range:
    0 - 40 mg/l NO₃-N, 0.01 - 0.40 mg/l NO₂- N
Specifications
About this item
Simple, low cost colour disc method of testing nitrate-nitrite.
Delivery: Includes colour disc, comparator box and tubes, reagents, instructions and carrying case.