Orders LinkContactUsLinkComponent
BinLiner™ Stands™
BinLiner™ Stands™
Supplier:   MICRONOVA
BinLiner™ Stands™
Supplier:  MICRONOVA
Certificates
BinLiner™ Stands™
BinLiner™ Stands™
4 Options Available Below
Product Details & Documents
The stainless steel Micronova BinLiner™ Stands™ are designed to work flawlessly with the BinLiners™ and similar sized bags. Securely hold a range of bin liner bags and feature two styles of open rod frames that allow ergonomic side removal of liners for simplified use.
  • Autoclavable and resistant to common cleaning solutions and disinfectants
The unique rectangular stainless steel wire stand includes plastic corners and feet to hold waste and garment disposal type bags/liners. The plastic feet (polyketone moulds) move easily on cleanroom floors without scratching, and eliminate rusting on floors from trash bag holders. Ergonomic side removal of liners simplifies use, compared to vertical removal from standard disposal bins. The corner legs are removable if disassembly is required. Available in a variety of sizes to hold 19 to 208 L bags.
The 'scissor' style stainless steel stand (BLSSH-40) easily folds for storage when not in use. Works best with 150 L bags.
Documents
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • MICRBLSSH-1515
    BinLiner stand, square, for 864×1016 mm and 813×914 mm bags
    BinLiner stand, square, for 864×1016 mm and 813×914 mm bags
    380×380×710 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 484,00 €
    484,00 €
    1 * 1 ST
       484,00 €
  • MICRBLSSH-168
    BinLiner stand, small rectangular, for 610×914 mm and 635×914 mm bags
    BinLiner stand, small rectangular, for 610×914 mm and 635×914 mm bags
    410×200×710 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 484,00 €
    484,00 €
    1 * 1 ST
       484,00 €
  • 129-1586
    BinLiner stand, large rectangular, for 1016×1219 mm and 1016×1524 mm bags
    BinLiner stand, large rectangular, for 1016×1219 mm and 1016×1524 mm bags
    610×410×860 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 484,00 €
    484,00 €
    1 * 1 ST
       484,00 €
  • 129-1585
    BinLiner stand, scissor style, for 1016×1219 mm bags
    BinLiner stand, scissor style, for 1016×1219 mm bags
    610×440×660 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 484,00 €
    484,00 €
    1 * 1 ST
       484,00 €