Calibration standards for osmometers
Supplier: GONOTEC
Certificates
Frequently Bought Together
Accessories for osmometer, Osmomat 3000
Supplier: GONOTEC
Decontamination Solution, DNA/RNA-ExitusPlus™ IF (Indicator-free)
Supplier: PanReac AppliChem
Ethanol denatured 96% ≥96%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium chloride 5 mol/L (5 M) in aqueous solution for molecular biology
Supplier: PanReac AppliChem
VWR®, Polyester Films for ELISA and General Incubation
Supplier: VWR Collection
Calibration standards for osmometers
Supplier: GONOTEC
Buffer solutions in sachets
Supplier: Mettler - Toledo
Accessories for pipette controller, BRAND accu-jet® S
Supplier: Brand
Storage and transport boxes, DURAporter™
Supplier: HEATHROW
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Total Organic Carbon (TOC) cleaning validation sets
Supplier: Texwipe
Murexide, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
Die-cut thermal transfer labels, B-7425
Supplier: Brady
Separating funnel stands, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF