Orders LinkContactUsLinkComponent
Sampling spoons, LaboPlast
Sampling spoons, LaboPlast
Supplier:   BURKLE
Sampling spoons, LaboPlast
Supplier:  BURKLE
Certificates
Sampling spoons, LaboPlast
Sampling spoons, LaboPlast
2 Options Available Below
Product Details & Documents
PS, white. These disposable spoons are ideal for sampling powders, granules and pastes.
  • Available with capacity of 2,5 ml (similar to teaspoon) and 10 ml (similar to tablespoon)
  • Production, assembling and packaging acc. to ISO cleanroom class 7
  • According to EU food and FDA guidelines
  • Individually packed
Product Family Options
Product Information
  • Length
    Capacity
    Availability
    Price
  • 127 mm
    2,5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 63,80 €
    63,80 €
    1 * 100 ST
       63,80 €
  • 170 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 68,00 €
    68,00 €
    1 * 100 ST
       68,00 €