Orders LinkContactUsLinkComponent
Sampling scoops, LaboPlast®
Sampling scoops, LaboPlast®
Supplier:   BURKLE
Sampling scoops, LaboPlast®
Supplier:  BURKLE
Certificates
Sampling scoops, LaboPlast®
Sampling scoops, LaboPlast®
14 Options Available Below
Product Details & Documents
PS, white. These scoops are ideal for the sampling of powder, granules and pastes. They are also available with a transparent lid to close it immediately after the sampling procedure to secure a clean and contamination-free sample.
  • Individually packed
  • Production, assembling and packaging acc. to ISO cleanroom class 7
  • According to EU food and FDA guidelines
Due to the special handle construction and the slightly angled edge the scoop stands horizontal and can be laid down without the material trickling out.
Product Family Options
Product Information
  • Type
    Capacity
    Length
    Colour
    Availability
    Price
  • Standard
    25 ml
    141 mm
    White
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 27,70 €
    27,70 €
    1 * 10 ST
       27,70 €
  • With lid
    25 ml
    142 mm
    White
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 43,20 €
    43,20 €
    1 * 10 ST
       43,20 €
  • Standard
    50 ml
    160 mm
    White
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 28,70 €
    28,70 €
    1 * 10 ST
       28,70 €
  • With lid
    50 ml
    161 mm
    White
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 44,70 €
    44,70 €
    1 * 10 ST
       44,70 €
  • Standard
    100 ml
    205 mm
    White
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 29,70 €
    29,70 €
    1 * 10 ST
       29,70 €
  • With lid
    100 ml
    206 mm
    White
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 45,90 €
    45,90 €
    1 * 10 ST
       45,90 €
  • Standard
    150 ml
    216 mm
    White
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 30,30 €
    30,30 €
    1 * 10 ST
       30,30 €
  • With lid
    150 ml
    218 mm
    White
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 46,80 €
    46,80 €
    1 * 10 ST
       46,80 €
  • Standard
    250 ml
    232 mm
    White
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 37,00 €
    37,00 €
    1 * 10 ST
       37,00 €
  • With lid
    250 ml
    235 mm
    White
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 62,70 €
    62,70 €
    1 * 10 ST
       62,70 €
  • Standard
    500 ml
    280 mm
    White
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 65,40 €
    65,40 €
    1 * 10 ST
       65,40 €
  • With lid
    500 ml
    283 mm
    White
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 112,00 €
    112,00 €
    1 * 10 ST
       112,00 €
  • Standard
    1000 ml
    332 mm
    White
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 84,80 €
    84,80 €
    1 * 10 ST
       84,80 €
  • Standard
    2000 ml
    381 mm
    White
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 97,10 €
    97,10 €
    1 * 10 ST
       97,10 €