Orders LinkContactUsLinkComponent
Pipette tips, epT.I.P.S.® Motion
Pipette tips, epT.I.P.S.® Motion
Supplier:   EPPENDORF
Pipette tips, epT.I.P.S.® Motion
Supplier:  EPPENDORF
Certificates
Pipette tips, epT.I.P.S.® Motion
Pipette tips, epT.I.P.S.® Motion
10 Options Available Below
Product Details & Documents
These pipette tips are specially designed for use on the epMotion 5070 and 5075 Liquid Handling Workstation. They were developed in torsion-resistant, contamination-free racks.
  • High-precision manufacturing guarantees a perfect fit and extremely accurate dispensing behavior
  • Tips are optimised for simple automatic ejection in the Liquid Handling Workstation collection vessel
  • Tips can also be integrated into the workstation as a refilling system
  • Refilling tips are delivered in a PET thermoformed tray with sealed lid
  • Replacing the disposable box with an autoclavable aluminum adapter can mean a waste savings of up to 40 percent
The epT.I.P.S. Motion Rack is made up of two components for flexible use in the automated epMotion Liquid Handling Workstation: a colour-coded tray with the epT.I.P.S. filter tips and a disposable box for storing the tray.
Packaging: Packaged in contamination-free racks or in reload trays.
Product Family Options
Product Information
  • Tip style
    Volume
    Sterile
    Packed
    Availability
    Price
  • Standard
    0,2 - 10 µl
    -
    Reloads, 24×96 tips
    Log in for availability
    Log in for availability
    1 * 2.304 ST
    1 * 2.304 ST - 258,00 €
    258,00 €
    1 * 2.304 ST
       258,00 €
  • Standard
    1 - 50 µl
    -
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 118,00 €
    118,00 €
    1 * 960 ST
       118,00 €
  • Standard
    1 - 50 µl
    -
    Reloads, 24×96 tips
    Log in for availability
    Log in for availability
    1 * 2.304 ST
    1 * 2.304 ST - 258,00 €
    258,00 €
    1 * 2.304 ST
       258,00 €
  • Standard
    20 - 300 µl
    -
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 118,00 €
    118,00 €
    1 * 960 ST
       118,00 €
  • Standard
    20 - 300 µl
    -
    Reloads, 24×96 tips
    Log in for availability
    Log in for availability
    1 * 2.304 ST
    1 * 2.304 ST - 258,00 €
    258,00 €
    1 * 2.304 ST
       258,00 €
  • Standard
    40 - 1000 µl
    -
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 118,00 €
    118,00 €
    1 * 960 ST
       118,00 €
  • Standard
    40 - 1000 µl
    -
    Reloads, 24×96 tips
    Log in for availability
    Log in for availability
    1 * 2.304 ST
    1 * 2.304 ST - 258,00 €
    258,00 €
    1 * 2.304 ST
       258,00 €
  • Standard
    1 - 50 µl
    +
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 136,00 €
    136,00 €
    1 * 960 ST
       136,00 €
  • Standard
    20 - 300 µl
    +
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 136,00 €
    136,00 €
    1 * 960 ST
       136,00 €
  • Standard
    40 - 1000 µl
    +
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 136,00 €
    136,00 €
    1 * 960 ST
       136,00 €