Orders LinkContactUsLinkComponent
Complement reagents
Complement reagents
Certificates
Complement reagents
1 Options Available Below
Product Details & Documents
Complement reagents are prepared from healthy, non-medicated animals using very large pools to reduce lot-to-lot variability. The fresh frozen complement is rapidly processed and maintained at –70 °C to ensure maximum activity. The lyophilised complement is supplied with a diluent of veronal buffered saline containing optimal concentrations of magnesium and calcium.
Specifications for Product Family
Specifications
  • Catalog No:
  • ROCKC300-0050
  • ROCKC300-0100
  • ROCKC300-0500
  • ROCKC301-0010
  • ROCKC301-0050
  • ROCKC301-0100
  • ROCKC302-0010
  • ROCKC303-0010
  • ROCKC303-0050
  • ROCKC304-0010
  • ROCKC304-0050
  • ROCKC304-0100
  • ROCKC304-0500
  • ROCKC305-0010
  • ROCKC305-0050
  • ROCKC305-0100
  • ROCKC306-0010
  • ROCKC306-0050
  • ROCKC306-0100
  • ROCKC309-0050
  • ROCKC309-0100
  • ROCKC200-0005
  • ROCKC201-0005
  • ROCKC202-0005
  • ROCKC203-0005
  • ROCKC204-0005
  • ROCKC205-0005
  • ROCKC206-0005
  • ROCKC209-0005
  • ROCKC220-0002
  • Formulation:
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
Specifications
Product Family Options
Lyophilised complement
Product Information
  • Availability
    Price
  • ROCKC200-0005
    Guinea pig complement
    Guinea pig complement
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC200-0005
    Formulation
    Lyophilized
  • ROCKC201-0005
    Mouse complement
    Mouse complement
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC201-0005
    Formulation
    Lyophilized
  • ROCKC202-0005
    Rabbit A (nesting) complement
    Rabbit A (nesting) complement
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC202-0005
    Formulation
    Lyophilized
  • ROCKC203-0005
    Rabbit B' (450 - 900 g) complement
    Rabbit B' (450 - 900 g) complement
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC203-0005
    Formulation
    Lyophilized
  • ROCKC204-0005
    Rabbit C (2265 - 2720 g) complement
    Rabbit C (2265 - 2720 g) complement
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC204-0005
    Formulation
    Lyophilized
  • ROCKC205-0005
    Rat complement
    Rat complement
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC205-0005
    Formulation
    Lyophilized
  • ROCKC206-0005
    Hamster complement
    Hamster complement
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC206-0005
    Formulation
    Lyophilized
  • ROCKC209-0005
    Goat complememnt
    Goat complememnt
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC209-0005
    Formulation
    Lyophilized
  • ROCKC220-0002
    Rabbit anti-Sheep cell hemolysin
    Rabbit anti-Sheep cell hemolysin
    Log in for availability
    Log in for availability
    1 * 2 mL
    1 * 2 mL - 441,00 €
    441,00 €
    1 * 2 mL
       441,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC220-0002
    Formulation
    Lyophilized
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.