Orders LinkContactUsLinkComponent
Blocking reagents, Blotto
Blocking reagents, Blotto
Blocking reagent, Blotto, immunoanalytical grade, 500 g
Catalog # ROCKB501-0500
Blocking reagents, Blotto
Catalog # ROCKB501-0500
CAS Number:  
Blocking reagents, Blotto
Blocking reagents, Blotto
Blocking reagent, Blotto, immunoanalytical grade, 500 g
CAS Number:  
Price
190,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Contains immunoanalytical grade non-fat dry milk. Blocks non specific binding of proteins in various immunological assays. Ideal for use in western blotting and ELISA methods.
Specifications
Specifications
  • Pk:
    500 g
  • Description:
    Blocking reagent, Blotto, immunoanalytical grade