Orders LinkContactUsLinkComponent
Blocking buffers
Blocking buffers
Blocking buffer for fluorescent western blotting, 500 mL
Catalog # ROCKMB-070
Blocking buffers
Catalog # ROCKMB-070
CAS Number:  
Blocking buffers
Blocking buffers
Blocking buffer for fluorescent western blotting, 500 mL
CAS Number:  
Price
337,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
A blocking buffer specifically designed for western blotting using fluorochrome conjugated antibodies. Blocking buffer was prepared using ultra pure reagents dissolved in pharmaceutical grade water (WFI) and consists of a proprietary protein formulation in tris buffered saline at pH 7,6 with thimerisol added as an antimicrobial agent.
Documents
Specifications
Specifications
  • Pk:
    500 mL
  • Description:
    Blocking buffer for fluorescent western blotting