Orders LinkContactUsLinkComponent
PCR plates, Eppendorf twin.tec® LoBind
PCR plates, Eppendorf twin.tec® LoBind
96, Standard, Fully skirted, 0,15 ml, Clear, Orange
  EPPE0030129.571
   Low Manufacturing Impact
 :   EPPENDORF
PCR plates, Eppendorf twin.tec® LoBind
  EPPE0030129.571
 :  EPPENDORF
   Low Manufacturing Impact
PCR plates, Eppendorf twin.tec® LoBind
PCR plates, Eppendorf twin.tec® LoBind
96, Standard, Fully skirted, 0,15 ml, Clear, Orange
1 892,00 €
1 * 300 ST
Eppendorf twin.tec® PCR plates combine extremely thin walled polypropylene wells for optimal heat transfer to the sample and robust polycarbonate frames for ultimate rigidity and torque-resistance. Eppendorf twin.tec® plates reduce the risk of cross contamination with raised well rims for effective sealing.
  • PCR clean
These Eppendorf twin.tec® 96 plates are ideal for quantitative Real-Time PCR, standard PCR, high-throughput PCR and small volume sample handling.
Ordering information: PCR plates are available with barcode upon request, please contact your local Avantor sales office for further details.
  • Frame colour:
    Orange
  • No. of wells:
    96
  • Profile:
    Standard
  • Skirt:
    Fully skirted
  • Well colour:
    Clear
  • Seq:
    10
  • Well volume:
    0,15 ml
  • Cat. No.:
    EPPE0030129.571
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact