Orders LinkContactUsLinkComponent
Gel drying Frames™
Gel drying Frames™
Gel drying Frames™
Gel drying Frames™
Gel drying Frames™
Gel drying frames allow researchers for easy handling and crack free dry gels without the use of heat/vacuum dryers.
Delivery information: The complete kit includes 2 open frames, 2 solid back plates, 12 side clips and 50 sheets of pre-cut cellophane.
Product Family Options
  • Description
  • Gel drying frame, 140×140 mm
    1 * 1 KIT
    1 * 1 KIT - 363,00 €
    363,00 €
    1 * 1 KIT
       363,00 €
  • Cellophane, 140×140 mm
    1 * 50 ST
    1 * 50 ST - 46,60 €
    46,60 €
    1 * 50 ST
       46,60 €