Water with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Certificates
About this item
:7732-18-5
:Packaged under nitrogen.
Storage Temperature:Ambient
EINECS:200-579-1
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Nominal concentration (V/V):0.095 - 0.105 %
- Evaporation residue:Max. 0.0001 %
- Ca (Calcium):Max. 0.2 ppm
- K (Potassium):Max. 0.2 ppm
- Mg (Magnesium):Max. 0.2 ppm
- Na (Sodium):Max. 0.5 ppm
- Transmittance (225 nm):Min. 20 %
- Transmittance (235 nm):Min. 50 %
- Transmittance (240 nm):Min. 80 %
- Transmittance (250 nm):Min. 95 %
- Transmittance (260 nm):Min. 99 %
- Suitable for LC-MS:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number Frequently Bought Together