Pipette tips, WHEATON®
Catalog # WHEA851358
Supplier: DWK Life Sciences
Specifications
- Packed:Case of 100 tips
- Sterile:+
- Volume:10 ml
- Seq:03
- Tip style:Macro
- Filter:+
- Pk:100
Specifications
About this item
These high quality tips are made of high grade, virgin, metal-free PP.
- Compatible with most macro pipettes
- Precision injection molding care for air-tight fit
- Free from trace metals for excellent accuracy and reproducibility