Order Entry
Belgium
ContactUsLinkComponent
Phenol red, BAKER ANALYZED® ACS indicator, J.T.Baker®
Phenol red, BAKER ANALYZED® ACS indicator, J.T.Baker®
Catalog # T254-01
Supplier:  Avantor
CAS Number:  
undefined
Phenol red, BAKER ANALYZED® ACS indicator, J.T.Baker®
Catalog # T254-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, wide mouth
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    Passes Test
  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Clarity of Solution:
    Passes Test
  • Visual Transition Interval - pH…6.8 (Yellow):
    Passes Test
  • Visual Transition Interval - pH…8.2 (Red):
    Passes Test

Specification Test Results

About this item

 :
Merck Index:13,07329
Melting Pt:>300 °C
 :354.38 g/mol
Storage Temperature:Ambient
EINECS:205-609-7
CAS Number:143-74-8
Formula:C₁₉H₁₄O₅S
MDL Number:MFCD00003552