Order Entry
Belgium
ContactUsLinkComponent
 
Tetrabutylammonium hydroxide 0.4 M in aqueous solution for HPLC, for ion pair chromatography, J.T.Baker®
  V365-07
 :  Avantor
 :  
undefined
Tetrabutylammonium hydroxide 0.4 M in aqueous solution for HPLC, for ion pair chromatography, J.T.Baker®
  V365-07
 :  Avantor
 :  

 

  • Pack type:
    HPPE poly bottle, pressure bottom
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • For Laboratory,Research,or Manufacturing Use:
    0.37 - 0.43
  • Molarity:
    0.37 - 0.43
  • Appearance:
    Passes Test
  • Absorbance at 420 nm (neat):
    ≤0.150
  • Bromide (Br):
    ≤0.05 %

 

 

 :
 :259.48 g/mol
 :Refrigerator
 :218-147-6
 :3267
 :8,II
 :2052-49-5
 :C₁₆H₃₇NO
 :MFCD00009425