Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 5 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Corrosive
Irritant
Sodium hydroxide 5 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Sodium hydroxide 5 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
  • Protect from air to avoid absorption of carbon dioxide.:
  • Appearance (Clear, colorless liquid):
    Passes Test
  • Normality:
    4.95 - 5.05
  • Chloride (Cl):
    ≤5 ppm
  • ACS - Heavy Metals (as Pb):
    ≤1 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.5 ppm
  • Standardization at 25°C traceable to NIST Standard Reference Material.:

Specification Test Results

Related Information

Frequently Bought Together