Order Entry
Belgium
ContactUsLinkComponent
Sodium metabisulphite NF, Multi-Compendial, J.T.Baker®
Sodium metabisulphite NF, Multi-Compendial, J.T.Baker®
  3551-05
 :  Avantor
 :  
undefined
Sodium metabisulphite NF, Multi-Compendial, J.T.Baker®
  3551-05
 :  Avantor
 :  

 

  • Pack type:
    Poly bottle, T/E cap
  • Pk:
    2,5 kg
  • Seq:
    4076

 

 

  • GMP Manufactured Product:
    Passes Test
  • Meets J.P. Chemical Specifications:
    Passes Test
  • Meets B.P. Chemical Specifications:
    = 0.05 %
  • Meets E.P. Chemical Specifications:
    = 0.002 %
  • Meets N.F. Requirements:
    = 0.05 %
  • CAUTION: For Manufacturing, processing or repackaging:
    = 0.002 %
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Limit of Chloride:
    ≤ 0.05 %
  • NF - Iron (Fe):
    ≤ 0.002 %
  • NF - Limit of Thiosulfate:
    ≤ 0.05 %
  • NF - Assay (SO₂):
    65.0 - 67.4 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Thiosulfate:
    Passes Test
  • EP/BP - Iron (Fe):
    ≤ 20 ppm
  • EP/BP - Assay (Na₂S₂O₅):
    95.0 - 100.5 %
  • EP/BP - pH:
    3.5 - 5.0
  • JP - Assay (Na₂S₂O₅):
    95.0 - 100.5 %
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Thiosulfate:
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤ 20 ppm
  • JP - Iron (Fe):
    ≤ 20 ppm
  • JP - Arsenic (As):
    ≤ 4 ppm
  • No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
    = 20 ppm

 

 

 :
 :190.11 g/mol
 :12,08784
 :150 °C
 :1.48 g/cm³ (20 °C)
 :Management of Change (MOC) category = R
 :231-673-0
 :01-2119531326-45
 :GHS05
 :GHS07
 :Ambient
 :7681-57-4
 :Na₂S₂O₅
 :MFCD00167602