Order Entry
Belgium
ContactUsLinkComponent
Sodium thiosulphate, anhydrous ≥98.0%, granular, BAKER ANALYZED®, J.T.Baker®
Sodium thiosulphate, anhydrous ≥98.0%, granular, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium thiosulphate, anhydrous ≥98.0%, granular, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 98.0 %
  • Assay (Na₂S₂O₃):
    ≥98.0 %
  • Insoluble Matter:
    ≤0.01 %
  • Calcium (Ca):
    ≤0.01 %
  • Heavy Metals (as Pb):
    ≤0.001 %
  • Sulfide (S):
    ≤1 ppm

Specification Test Results

Related Information

Frequently Bought Together