Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid 95.0-98.0% NF, Multi-Compendial, J.T.Baker®
Corrosive
Irritant
Health
Sulphuric acid 95.0-98.0% NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Health
Sulphuric acid 95.0-98.0% NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    Passes Test
  • NF - Identification:
    Passes Test
  • NF - Residue on Ignition:
    ≤ 0.005 %
  • NF - Chloride (Cl):
    ≤ 0.005 %
  • NF - Arsenic (As):
    ≤ 1 ppm
  • NF - Reducing Substances:
    Passes Test
  • NF - Assay (as H₂SO₄) (by acidimetry):
    95.0 - 98.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤ 50 ppm
  • EP/BP - Nitrate (NO₃):
    Passes Test
  • EP/BP - Iron (Fe):
    ≤ 25 ppm
  • EP/BP - Assay (H₂SO₄):
    95.0 - 100.5 %
  • No Class 1,2,3 or other solvents are used or produced in the:
    = 5 ppm
  • manufacturing or purification of the product.:
    95.0 - 100.5 %

Specification Test Results

Related Information

Frequently Bought Together