Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Centrifuges
Rotors
Rotor for ROTO SILENTA 630 RS centrifuge
Rotor for ROTO SILENTA 630 RS centrifuge
Rotor for ROTO SILENTA 630 RS centrifuge
Catalog # HETT4176
Supplier:
Hettich
Expand image
Rotor for ROTO SILENTA 630 RS centrifuge
Catalog # HETT4176
Supplier:
Hettich
Specifications
Description:
Swing out rotor without carriers, for 6 tubes and bottles up to 2000 ml, 4500 min
-1
, 6500 ×g
Seq:
01
Pk:
1
Specifications
About this item
Product Details
Product Details