Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
Conductivity standards
Supplier:  HAMILTON BONADUZ
Certificates
Conductivity standards
Conductivity standards
8 Options Available Below
Product Details & Documents
Conductivity standards with stability of ±1% - guaranteed over a lifetime of up to three years.
  • Remains stable for a minimum of 1 year for 1,3 μs/cm, and up to 3 years for all other values
  • Certificate with calibration document from DFM
  • Expiration date shown on every bottle
  • Bottles are permitted to stay open for a total of 60 minutes
The procedure for determining conductivity was developed in collaboration with DFM (Danish Institute of Fundamental metrology). A representative number of bottles from every batch are measured by DFM. Their value is recorded on the calibration certificate and on every bottle. DFM enjoys the highest prestige in Europe in the area of electrolytic conductivity and is equipped with an absolute measurement cell that was developed in collaboration with NIST, and is accredited by the Danish accreditation agency DANAK to a conductivity of 0,9 μs/cm.
Product Family Options
Product Information
  • Conductivity
    Pk
    Availability
    Price
  • HAMI238926
    Conductivity standard in glass bottle, with 36 months stability
    Conductivity standard in glass bottle, with 36 months stability
    5 µS/cm
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 149,00 €
    149,00 €
    1 * 250 mL
       149,00 €
  • HAMI238988
    Conductivity standard, Basic Line, in Calpack bottle, with 18 months stability
    Conductivity standard, Basic Line, in Calpack bottle, with 18 months stability
    12 880 µS/cm
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 67,50 €
    67,50 €
    1 * 500 mL
       67,50 €
  • HAMI238927_U
    Conductivity standard in glass bottle, with 36 months stability
    Conductivity standard in glass bottle, with 36 months stability
    15 µS/cm
    300 mL
    Log in for availability
    Log in for availability
    1 * 300 mL
    1 * 300 mL
  • HAMI238984
    Conductivity standard, Basic Line, in Calpack bottle, with 18 months stability
    Conductivity standard, Basic Line, in Calpack bottle, with 18 months stability
    84 µS/cm
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 67,50 €
    67,50 €
    1 * 500 mL
       67,50 €
  • HAMI238985
    Conductivity standard, Basic Line, in Calpack bottle, with 18 months stability
    Conductivity standard, Basic Line, in Calpack bottle, with 18 months stability
    147 µS/cm
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 67,50 €
    67,50 €
    1 * 500 mL
       67,50 €
  • HAMI238929
    Conductivity standard in glass bottle, with 36 months stability
    Conductivity standard in glass bottle, with 36 months stability
    706 µS/cm
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 125,00 €
    125,00 €
    1 * 250 mL
       125,00 €
  • HAMI238928
    Conductivity standard in glass bottle, with 36 months stability
    Conductivity standard in glass bottle, with 36 months stability
    1413 µS/cm
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 93,80 €
    93,80 €
    1 * 250 mL
       93,80 €
  • HAMI238986
    Conductivity standard, Basic Line, in Calpack bottle, with 18 months stability
    Conductivity standard, Basic Line, in Calpack bottle, with 18 months stability
    1413 µS/cm
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 67,50 €
    67,50 €
    1 * 500 mL
       67,50 €