Orders LinkContactUsLinkComponent
Isolator cleaning tool
Supplier:   Texwipe
Isolator cleaning tool
Supplier:  Texwipe
Certificates
Isolator cleaning tool
Isolator cleaning tool
6 Options Available Below
Product Details & Documents
Isolator cleaning tool: Mop hardware, replacement head, replacement handle.
Specially engineered to work with isolators and other small areas, this easy to manoeuvre tool offers lightweight, one handed operation. With a low profile, swivel head, the isolator cleaning tool gets into 'hard to reach' places.
Engineered for quick cover changes during operation, the padded head allows for a consistent mop across different surfaces without losing cleaning power. The rounded edges clean the isolator corners and prevent scratches or damage to surfaces.
  • Rapid change system allows user to replace covers/pads frequently, adhering to strict cleaning protocols
  • Polyester pad conforms to surfaces
  • Gets into 'hard to reach' areas with its low profile, swivel head
  • 100% polyester covers provide the cleanest substrates available for cleaning isolators
  • Offers one handed operation with lightweight handles
Isolator cleaning covers:
Made from 100% polyester or 100% polyester microdenier fabric. This fabric provides enhanced particle lifting, streak-free cleaning, plus enhanced sorption for working with solutions.
Enhanced absorbency and quick-wicking performance
Increased lifting and capturing of particles as small as microbes Streak-free cleaning.
Made from 100% polyester or 100% polyester microdenier fabric. This fabric provides enhanced particle lifting, streak-free cleaning, plus enhanced sorption for working with solutions.
Product Family Options
Replacement head
Product Information
  • L×W
    Length
    Packed
    Availability
    Price
  • 141-1054
    Isolator cleaning tool replacement head: with 1 thermoplastic mop head (100×180 mm), 1 pad
    Isolator cleaning tool replacement head: with 1 thermoplastic mop head (100×180 mm), 1 pad
    100×180 mm
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 121,66 €
    121,66 €
    1 * 1 ST
       121,66 €
Replacement handles
Product Information
  • L×W
    Length
    Packed
    Availability
    Price
  • 141-1061
    Isolator cleaning tool replacement handle, white fibreglass, autoclave safe
    Isolator cleaning tool replacement handle, white fibreglass, autoclave safe
    304 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,64 €
    27,64 €
    1 * 1 ST
       27,64 €
  • 141-1062
    Isolator cleaning tool replacement handle, white fibreglass, autoclave safe
    Isolator cleaning tool replacement handle, white fibreglass, autoclave safe
    450 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,25 €
    30,25 €
    1 * 1 ST
       30,25 €
  • 141-1063
    Isolator cleaning tool extension handle telescoping, white fibreglass, autoclave safe
    Isolator cleaning tool extension handle telescoping, white fibreglass, autoclave safe
    730 - 1340 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,52 €
    77,52 €
    1 * 1 ST
       77,52 €
Isolator cleaning covers
Product Information
  • L×W
    Length
    Packed
    Availability
    Price
  • 141-1047
    Sterile isolator cleaning tool, microdenier covers and polyester pads
    Sterile isolator cleaning tool, microdenier covers and polyester pads
    5 covers and 1 pad/ bag; 25 bags/case
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 726,83 €
    726,83 €
    1 * 1 KIT
       726,83 €
  • TEXWSTX7114
    Sterile Isolator cleaning tool polyester covers and polyester pads
    Sterile Isolator cleaning tool polyester covers and polyester pads
    5 covers and 1 pad/ bag; 25 bags/case
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 513,79 €
    513,79 €
    1 * 1 KIT
       513,79 €
    Specifications:
    New Product