ContactUsLinkComponent
Sterile anti-fog solution for autoclavable goggles, BioClean™
Sterile anti-fog solution for autoclavable goggles, BioClean™
Catalog # 113-8145
Supplier:  Nitritex
undefined
Sterile anti-fog solution for autoclavable goggles, BioClean™
Catalog # 113-8145
Supplier:  Nitritex
Specifications
  • Description:
    Sterile anti-fog solution for autoclavable goggles, BioClean, 60 ml bottle
  • Size:
    60 ml
  • Pk:
    10
Specifications
About this item
Sterile anti-fog spray for autoclavable goggles.
  • Only two sprays to be applied on the inside of the goggles
  • Excess liquid from the inside to be dried by cleanroom wipe
  • To be used after each autoclave cycle
  • Sterilised by gamma irradiation