Order Entry
Belgium
ContactUsLinkComponent
Hexane (mixture of isomers) ≥98.5% (by GC, sum of enantiomers), BAKER ANALYZED® ACS, J.T.Baker®
Flammable
Health
Irritant
Environment
Hexane (mixture of isomers) ≥98.5% (by GC, sum of enantiomers), BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Flammable
Health
Irritant
Environment
Hexane (mixture of isomers) ≥98.5% (by GC, sum of enantiomers), BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    = 98.5 %
  • ACS - Assay (Sum of 5 isomers, total hexanes plus methylcyclopentane) (by GC):
    ≥ 98.5 %
  • ACS - Color (APHA):
    ≤ 10
  • ACS - Residue after Evaporation:
    ≤ 0.001 %
  • ACS - Water-Soluble Titrable Acid, meq/g:
    ≤ 0.0003
  • ACS - Sulfur Compounds (as S):
    ≤ 0.005 %
  • ACS - Thiophene:
    Passes Test
  • Product Information (not specifications)::
    68.7 °C
  • Boiling Point (typical)................. 68.7 °C.:
    0.663

Specification Test Results

Related Information

Frequently Bought Together