Orders LinkContactUsLinkComponent
Stopcocks, one-way
Stopcocks, one-way
Female Luer to Female Luer
Catalog # POPR6020
Supplier:   CADENCE SCIENCE
Stopcocks, one-way
Catalog # POPR6020
Supplier:  CADENCE SCIENCE
Stopcocks, one-way
Female Luer to Female Luer
Price
321,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents
Theses stopcocks, made of nickel plated brass, are precision-machined, individually lapped, and carefully mated to maintain pressure characteristics. These stopcocks are easily integrated with other stopcocks, manifolds, adapters, tubing, needles, and glass syringes.
Specifications
  • Description:
    Female Luer to Female Luer
  • Seq:
    02