Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Weights
Calibration Weights
VWR®, Calibration Weight Sets, Class M1
VWR®, Calibration Weight Sets, Class M1
VWR®, Calibration Weight Sets, Class M1
611-1854
:
VWR Collection
VWR®, Calibration Weight Sets, Class M1
611-1854
:
VWR Collection
Weight set:
10 g; 50 g; 100 g
Pk:
1
No. of weights:
3
10 g stainless steel knob weight
50 g stainless steel knob weight
100 g nickel plated brass knob weight
:
Weights are supplied in a bag.