ContactUsLinkComponent
Immobilised Pepstatin for purification of Cathepsins/Pepsin/Pepstatin binding molecules
Immobilised Pepstatin for purification of Cathepsins/Pepsin/Pepstatin binding molecules
Catalog # 786-789
Supplier:  G-Biosciences
CAS Number:  
undefined
Immobilised Pepstatin for purification of Cathepsins/Pepsin/Pepstatin binding molecules
Catalog # 786-789
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    5 mL
  • Description:
    Immobilised Pepstatin
Specifications
About this item
Immobilised Pepstatin is for the purification of cathepsins, pepsin, bacterial aspartic proteases, HIV proteases and other molecules that bind pepstatin.