Carbohydrate coupling resin
About this item
Carbohydrate coupling resin is designed for the covalent immobilisation of carbohydrate containing molecules, including glycoproteins, to agarose beads. It is ideal for the coupling of polyclonal antibodies as it allows for the optimal orientation of the antibodies for affinity purification.
The Carbohydrate Immobilization Kit is supplied with carbohydrate coupling resin columns for the preparation of 2 ml affinity columns, a coupling catalyst, SpinOUT™ GT-600 Desalting Columns for removing oxidising reagents, the oxidizing reagent (sodium meta-periodate) and relevant buffers.
2 Options Available Below
Product Details & Documents
Carbohydrate coupling resin is designed for the covalent immobilisation of carbohydrate containing molecules, including glycoproteins, to agarose beads. It is ideal for the coupling of polyclonal antibodies as it allows for the optimal orientation of the antibodies for affinity purification.
The Carbohydrate Immobilization Kit is supplied with carbohydrate coupling resin columns for the preparation of 2 ml affinity columns, a coupling catalyst, SpinOUT™ GT-600 Desalting Columns for removing oxidising reagents, the oxidizing reagent (sodium meta-periodate) and relevant buffers.
Documents
GENO786-808_TDS_EN
Datasheet
Product Family Options
Product Information