Orders LinkContactUsLinkComponent
Protein reducing agents
Protein reducing agents
Supplier:   G-Biosciences
Protein reducing agents
Supplier:  G-Biosciences
Certificates
Protein reducing agents
Protein reducing agents
2 Options Available Below
Product Details & Documents
Protein-S-S-Reductant™ uses TCEP (Tris [2-carboxyethyl] phosphine), a popular alternative to β-mercaptoethanol and DTT (dithiothreitol). TCEP improves stability, increases effectiveness, and reduces proteins over a wider range of pH, including lower acidic pHs. Protein-S-S-Reductant™ completely reduces stable disulfide bonds in less than 5 minutes at room temperature and is compatible with the protein alkylation reactions. This ready to use solution is at a neutral pH and stabilised for long-term storage. Simply supplement Protein-S-S-Reductant™ in place of DTT or β-mercaptoethanol and boil the sample.
OneQuant™ DTT are single aliquots of DTT that eliminate the need for weighing; preventing loss of reagent and saving time. Add 90 μl water to a single tube to generate a 0,5 M DTT solution. Supplied with 40 individual tubes.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 786-25PR
    Protein-S-S-Reductant™
    Protein-S-S-Reductant™
    200 Tests
    Log in for availability
    Log in for availability
    1 * 200 Tests
    1 * 200 Tests - 178,00 €
    178,00 €
    1 * 200 Tests
       178,00 €
  • 786-077
    OneQuant™ DTT, 0,5 M
    OneQuant™ DTT, 0,5 M
    40
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 253,00 €
    253,00 €
    1 * 40 ST
       253,00 €