ContactUsLinkComponent
CDI amine reactive resin
CDI amine reactive resin
Catalog # 786-404
Supplier:  G-Biosciences
CAS Number:  
undefined
CDI amine reactive resin
Catalog # 786-404
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    10 mL
  • Description:
    CDI amine reactive resin
Specifications
About this item
CDI amine reactive agarose consists of 6% cross-linked agarose activated with CDI (1,1’-carbonyl diimidazole) to form reactive imidazole carbamates.
  • Proven coupling chemistry and stable linkages
  • Easy to use
  • Couple in inorganic buffers for insoluble molecules
  • Couple proteins and peptides
  • Couple primary amine containing ligands
Ideal for immobilising peptides, small organic molecules and certain proteins and reactions can occur in organic solvent making it suitable for water-insoluble ligands.