Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bottles
Aspirators
Replacement cap for aspirator bottles
Replacement cap for aspirator bottles
Replacement cap for aspirator bottles
Catalog # 215-0093
Supplier:
SciLabware
Expand image
Replacement cap for aspirator bottles
Catalog # 215-0093
Supplier:
SciLabware
Specifications
Description:
Spare cap for BNP10A/B & 20A/B bottles (10 / 20 L)
Pk:
1
Specifications
About this item
Product Details
Product Details