Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Hydrobromic acid 47%, NORMATOM®, Ultrapure for trace metal analysis
  85032.270
 :  
undefined
 
 
Hydrobromic acid 47%, NORMATOM®, Ultrapure for trace metal analysis
  85032.270
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Assay:
    44-49%
  • Ag (Silver):
    100 ppt
  • Al (Aluminium):
    100 ppt
  • Au (Gold):
    100 ppt
  • B (Boron):
    Information only
  • Ba (Barium):
    100 ppt
  • Be (Beryllium):
    100 ppt
  • Bi (Bismuth):
    100 ppt
  • Ca (Calcium):
    100 ppt
  • Cd (Cadmium):
    100 ppt
  • Ce (Cerium):
    10 ppt
  • Co (Cobalt):
    100 ppt
  • Cr (Chromium):
    100 ppt
  • Cs (Cesium):
    10 ppt
  • Cu (Copper):
    100 ppt
  • Dy (Dysprosium):
    10 ppt
  • Er (Erbium):
    10 ppt
  • Eu (Europium):
    10 ppt
  • Fe (Iron):
    100 ppt
  • Ga (Gallium):
    100 ppt
  • Gd (Gadolinium):
    10 ppt
  • Hf (Hafnium):
    10 ppt
  • Ho (Holmium):
    10 ppt
  • In (Indium):
    10 ppt
  • K (Potassium):
    100 ppt
  • La (Lanthanum):
    10 ppt
  • Li (Lithium):
    100 ppt
  • Lu (Lutetium):
    10 ppt
  • Mg (Magnesium):
    100 ppt
  • Mn (Manganese):
    100 ppt
  • Mo (Molybdenum):
    100 ppt
  • Na (Sodium):
    100 ppt
  • Nb (Niobium):
    10 ppt
  • Nd (Neodymium):
    10 ppt
  • Ni (Nickel):
    100 ppt
  • Pb (Lead):
    1 ppt
  • Pd (Palladium):
    100 ppt
  • Pr (Praseodymium):
    10 ppt
  • Pt (Platinum):
    100 ppt
  • Rb (Rubidium):
    100 ppt
  • Re (Rhenium):
    100 ppt
  • Rh (Rhodium):
    100 ppt
  • Ru (Ruthenium):
    100 ppt
  • Sb (Antimony):
    Information only
  • Sc (Scandium):
    100 ppt
  • Sm (Samarium):
    10 ppt
  • Sn (Tin):
    100 ppt
  • Sr (Strontium):
    100 ppt
  • Ta (Tantalum):
    Information only
  • Tb (Terbium):
    10 ppt
  • Te (Tellurium):
    10 ppt
  • Th (Thorium):
    1 ppt
  • Ti (Titanium):
    100 ppt
  • Tl (Thallium):
    10 ppt
  • Tm (Thulium):
    10 ppt
  • U (Uranium):
    1 ppt
  • V (Vanadium):
    100 ppt
  • W (Tungsten):
    100 ppt
  • Y (Yttrium):
    10 ppt
  • Yb (Ytterbium):
    10 ppt
  • Zn (Zinc):
    100 ppt
  • Zr (Zirconium):
    10 ppt

 

 

 :
 :80.91 g/mol
 :1.47 g/cm³ (20 °C)
 :Ambient
 :233-113-0
 :1788
 :8,II
 :GHS05
 :GHS07
 :10035-10-6
 :HBr
 :MFCD00011323