Order Entry
Belgium
ContactUsLinkComponent
Hydrofluoric acid 47%, NORMATOM®, Ultrapure for trace metal analysis
Hydrofluoric acid 47%, NORMATOM®, Ultrapure for trace metal analysis
Supplier:  VWR Chemicals
Certificates
undefined
Hydrofluoric acid 47%, NORMATOM®, Ultrapure for trace metal analysis
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay:
    47-51%
  • Total S (Sulphur):
    100 ppb
  • Ag (Silver):
    10 ppt
  • Al (Aluminium):
    20 ppt
  • As (Arsenic):
    50 ppt
  • Au (Gold):
    20 ppt
  • B (Boron):
    100 ppt
  • Ba (Barium):
    10 ppt
  • Be (Beryllium):
    10 ppt
  • Bi (Bismuth):
    10 ppt
  • Ca (Calcium):
    10 ppt
  • Cd (Cadmium):
    10 ppt
  • Ce (Cerium):
    10 ppt
  • Co (Cobalt):
    10 ppt
  • Cr (Chromium):
    10 ppt
  • Cs (Cesium):
    10 ppt
  • Cu (Copper):
    10 ppt
  • Dy (Dysprosium):
    1 ppt
  • Er (Erbium):
    1 ppt
  • Eu (Europium):
    1 ppt
  • Fe (Iron):
    10 ppt
  • Ga (Gallium):
    10 ppt
  • Gd (Gadolinium):
    1 ppt
  • Ge (Germanium):
    10 ppt
  • Hf (Hafnium):
    10 ppt
  • Hg (Mercury):
    50 ppt
  • Ho (Holmium):
    1 ppt
  • In (Indium):
    1 ppt
  • K (Potassium):
    10 ppt
  • La (Lanthanum):
    10 ppt
  • Li (Lithium):
    10 ppt
  • Lu (Lutetium):
    1 ppt
  • Mg (Magnesium):
    10 ppt
  • Mn (Manganese):
    10 ppt
  • Mo (Molybdenum):
    10 ppt
  • Na (Sodium):
    10 ppt
  • Nb (Niobium):
    10 ppt
  • Nd (Neodymium):
    1 ppt
  • Ni (Nickel):
    20 ppt
  • Pb (Lead):
    10 ppt
  • Pd (Palladium):
    20 ppt
  • Pr (Praseodymium):
    1 ppt
  • Pt (Platinum):
    20 ppt
  • Rb (Rubidium):
    20 ppt
  • Re (Rhenium):
    10 ppt
  • Rh (Rhodium):
    20 ppt
  • Ru (Ruthenium):
    20 ppt
  • Sb (Antimony):
    20 ppt
  • Sc (Scandium):
    10 ppt
  • Se (Selenium):
    Information only
  • Sm (Samarium):
    1 ppt
  • Sn (Tin):
    20 ppt
  • Sr (Strontium):
    10 ppt
  • Ta (Tantalum):
    Information only
  • Tb (Terbium):
    1 ppt
  • Te (Tellurium):
    1 ppt
  • Th (Thorium):
    1 ppt
  • Ti (Titanium):
    20 ppt
  • Tl (Thallium):
    10 ppt
  • Tm (Thulium):
    1 ppt
  • U (Uranium):
    1 ppt
  • V (Vanadium):
    10 ppt
  • W (Tungsten):
    20 ppt
  • Y (Yttrium):
    1 ppt
  • Yb (Ytterbium):
    1 ppt
  • Zn (Zinc):
    10 ppt
  • Zr (Zirconium):
    10 ppt

Specification Test Results

Frequently Bought Together