Order Entry
Belgium
ContactUsLinkComponent
 
Ethidium bromide 0.625 mg/ml in aqueous solution
Catalog # R041
Supplier:  G-Biosciences
CAS Number:  
Ethidium bromide 0.625 mg/ml in aqueous solution
Catalog # R041
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :394.31 g/mol
CAS Number:1239-45-8
Formula:C₂₁H₂₀BrN₃
MDL Number:MFCD00011724