Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Ammonium nitrate ≥95.0%, crystals, AR® ACS, Macron Fine Chemicals™
 
undefined
 
 
Ammonium nitrate ≥95.0%, crystals, AR® ACS, Macron Fine Chemicals™
 

 

  • Meets ACS Reagent Chemical Requirements:
    = 95.0 %
  • For Laboratory,Research,or Manufacturing Use:
    = 5 ppm
  • ACS - Assay (as NH₄NO₃):
    ≥ 95.0 %
  • ACS - Chloride (Cl):
    ≤ 5 ppm
  • ACS - Heavy Metals (as Pb):
    ≤ 5 ppm
  • ACS - Insoluble Matter:
    ≤ 0.005 %
  • ACS - Iron (Fe):
    ≤ 2 ppm
  • ACS - Nitrite (NO₂):
    Passes Test
  • ACS - pH of 5% Solution at 25°C:
    4.5 - 6.0
  • ACS - Phosphate (PO₄):
    ≤ 5 ppm
  • ACS - Residue after Ignition:
    ≤ 0.01 %
  • ACS - Sulfate (SO₄):
    ≤ 0.002 %

 

 

Frequently Bought Together