Dry buffer packs, JAW™
Supplier: G-Biosciences
Jump to:
About this item
Premixed buffer powders that simply require adding water to generate a ready-to-use buffer.
4 Options Available Below
Product Details & Documents
Premixed buffer powders that simply require adding water to generate a ready-to-use buffer.
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- ContentspH valuePkAvailabilityPrice
- 786-289Phosphate buffered saline packs, JAW™, 1X, 1 l/packPhosphate buffered saline packs, JAW™, 1X, 1 l/pack2,7 mM potassium chloride, 137 mM sodium chloride and 10 mM phosphate buffer7,3 - 7,520Log in for availability1 * 20 ST130,00 €
- 786-288Tris buffered saline packs, JAW™, 1X, 1 l/packTris buffered saline packs, JAW™, 1X, 1 l/pack25mM Tris, 140mM sodium chloride and 3mM potassium chloride7,25 - 7,5520Log in for availability1 * 20 ST130,00 €
- RC-147Phosphate buffered saline (PBS), JAW™, 10X, dry buffer pack for 10 l of 1X/ packPhosphate buffered saline (PBS), JAW™, 10X, dry buffer pack for 10 l of 1X/ pack43 mM NaH₂PO₄, 1,37 M NaCl, 27mM KCl and 14 mM KH₂PO₄7,42Log in for availability1 * 2 ST130,00 €
- RC-148Tris buffered saline (TBS), JAW™, 20X, dry buffer pack for 20 l of 1X/ packTris buffered saline (TBS), JAW™, 20X, dry buffer pack for 20 l of 1X/ pack0,5 M Tris, 2,8 M sodium chloride and 60 mM potassium chloride7,25 - 7,551Log in for availability1 * 1 ST111,00 €