ContactUsLinkComponent
Phosphate buffered saline (PBS), 10X
Phosphate buffered saline (PBS), 10X
Catalog # 786-027
Supplier:  G-Biosciences
CAS Number:  
undefined
Phosphate buffered saline (PBS), 10X
Catalog # 786-027
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    500 mL
  • Description:
    PBS, 10×
Specifications
About this item
PBS 10X solution is used as a wash buffers for Western blotting, ELISA and other applications.