Orders LinkContactUsLinkComponent
Abbe refractometer
Abbe refractometer
Abbe refractometer, Novex, 0 - 95 °Bx, 1,30 - 1,70 nD, 0,5 °Bx; 0,0002 nD
N° de catalogue EUMX98.490
Fournisseur:   EUROMEX
Abbe refractometer
N° de catalogue EUMX98.490
Fournisseur:  EUROMEX
Abbe refractometer
Abbe refractometer
Abbe refractometer, Novex, 0 - 95 °Bx, 1,30 - 1,70 nD, 0,5 °Bx; 0,0002 nD
Prix
1 820,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
The Abbe laboratory refractometer is a bench-top instrument for high-precision measurements of an index of refraction.
  • More accurate than hand refractometers
  • Capable of measuring all kinds of concentrations and identifying several types of substances
It can be connected to a water bath for measuring at a controlled temperature and used for determining the refractive index of glass, plastic films. During operating this type of refractometer, the use of a cold light source is recommended. The instrument is equipped with a built-in thermometer and water connection to control fluid temperatures.
This Abbe refractometer has scales for measurement of sugar concentrations and refractive indices.
The use of a cold light source like the LE.5209 is recommended for this type of refractometer.
Livraison: Supplied with flight case, thermometer, test plate and adjusting tools.
Spécifications
  • Model:
    Novex
  • Range:
    0 - 95 °Bx
  • Resolution:
    0,5 °Bx; 0,0002 nD
  • Description:
    Abbe refractometer
  • Refractive index:
    1,30 - 1,70 nD
Accessories
Informations sur le produit
  • N° de catalogue
    Disponibilité
    Prix
  • Cold light source
    Cold light source
    EUMXLE.5209
    Connexion Pour la disponibilité
    1 * 1 ST