ContactUsLinkComponent
Apoptotic DNA ladder kit
Apoptotic DNA ladder kit
Catalog # 786-209
Supplier:  G-Biosciences
CAS Number:  
undefined
Apoptotic DNA ladder kit
Catalog # 786-209
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Apoptotic DNA Ladder Kit
Specifications
About this item
The Apoptotic DNA Ladder Kit contains the reagents necessary for the preparation of cellular fractions for the production of apoptosis nucleosomal DNA ladders. The kit does not require toxic phenol and DNA ladder is prepared either with or without genomic DNA.