Orders LinkContactUsLinkComponent
Apoptosis inducer set
Apoptosis inducer set
Apoptotic Inducer Set (Set of 6), 1 SET
Catalog # API-16S
Supplier:   G-Biosciences
Apoptosis inducer set
Catalog # API-16S
Supplier:  G-Biosciences
CAS Number:  
Apoptosis inducer set
Apoptosis inducer set
Apoptotic Inducer Set (Set of 6), 1 SET
CAS Number:  
Price
735,00 €
Connexion Pour la disponibilité
1 * 1 SET
Product Details & Documents
Apoptosis inducers are used for induction of apoptosis in cultured cells. The selection of apoptosis inducers include actinomycin D, camptothecin, cycloheximide, dexamethasone, doxorubicin and etoposide.
  • Ready to use
  • Long storage life
Documents
Spécifications
Spécifications
  • Pk:
    1 SET
  • Description:
    Apoptotic Inducer Set (Set of 6)
Spécifications