Destain solutions
Supplier: G-Biosciences
Certificates
About this item
G-Biosciences' Destain I contains approximately 45% methanol and 9% glacial acetic acid. G-Biosciences' Destain II contains approximately 5% methanol and 7% acetic acid.
Frequently Bought Together