Order Entry
Belgium
ContactUsLinkComponent
Ammonium nitrate ≥95.0%, BAKER ANALYZED® ACS, J.T.Baker®
Oxidizing
Irritant
Ammonium nitrate ≥95.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Oxidizing
Irritant
Ammonium nitrate ≥95.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    ≥95.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.005%
  • ACS - Assay (NH₄NO₃):
    ≥95.0 %
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Residue after Ignition:
    ≤0.01 %
  • ACS - pH of 5% Solution at 25°C:
    4.5 - 6.0
  • ACS - Sulfate (SO₄):
    ≤0.002 %
  • Chloride (Cl):
    ≤5 ppm
  • Nitrite (NO₂):
    Passes Test
  • Phosphate (PO₄):
    ≤5 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤2.0 ppm

Specification Test Results

Related Information

Frequently Bought Together