Order Entry
Belgium
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS) 20% in aqueous solution
Sodium dodecyl sulphate (SDS) 20% in aqueous solution
Catalog # 786-017
Supplier:  G-Biosciences
CAS Number:  
undefined
Sodium dodecyl sulphate (SDS) 20% in aqueous solution
Catalog # 786-017
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :288.38 g/mol
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175