ContactUsLinkComponent
Protease inhibitor set
Protease inhibitor set
Catalog # 786-207
Supplier:  G-Biosciences
CAS Number:  
undefined
Protease inhibitor set
Catalog # 786-207
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    1 SET
  • Description:
    Protease Inhibitor Set
  • Seq:
    02
Specifications
About this item
Protease inhibitor set contains 12 ready to use individual protease inhibitors, each at a 100X concentration. Various concentrations and/or combinations of protease inhibitors may be used to inhibit a broad spectrum of protease activity. The Protease inhibitor set can be used to design specific protease inhibitor cocktails, supplement existing cocktails or to screen for specific protease classes.
Delivery: Set includes each of the following individual protease inhibitors: AEBSF, ALLN, Antipain dihydrochloride, Aprotinin, Bestatin, Chymostatin, E-64, EDTA-Na₂, Leupeptin, Pepstatin, Phosphoramidon, and PMSF. Individual proteases are also available as dry powders.