Orders LinkContactUsLinkComponent
Biotin kits, Micro HOOK™
Biotin kits, Micro HOOK™
Supplier:   G-Biosciences
Biotin kits, Micro HOOK™
Supplier:  G-Biosciences
Certificates
Biotin kits, Micro HOOK™
Biotin kits, Micro HOOK™
4 Options Available Below
Product Details & Documents
The micro HOOK™ Biotin kits are designed to label small amounts of proteins, with each kit designed for 8 to 10 labellings of 50 to 250 µg protein/reaction.
  • Micro kit for labelling protein primary amines
  • Optimizer Buffer™ for improved coupling efficiency
  • Gel filtration columns for rapid (<10 minute) purification
  • Labels 50 to 250µg protein/reaction
  • Suitable for 8 to 10 couplings
Kits contain the following Biotin reagents
HOOK™ Sulfo-NHS-Biotin - amine reactive reagent, shortest spacer arm
HOOK™ Sulfo-NHS-LC-Biotin - amine reactive reagent, longer spacer arm
HOOK™ Sulfo-NHS-SS-Biotin - cleavable, amine reactive reagent
HOOK™ NHS-dPEG₄-Biotin - amine reactive pegylated reagent
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 786-694
    HOOK™-sulfo-NHS-Biotin Kit
    HOOK™-sulfo-NHS-Biotin Kit
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 457,00 €
    457,00 €
    1 * 1 KIT
       457,00 €
  • 786-695
    HOOK™-sulfo-NHS-LC-Biotin Kit
    HOOK™-sulfo-NHS-LC-Biotin Kit
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 483,00 €
    483,00 €
    1 * 1 KIT
       483,00 €
  • 786-696
    HOOK™ Sulfo-NHS-SS-Biotin Kit
    HOOK™ Sulfo-NHS-SS-Biotin Kit
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 483,00 €
    483,00 €
    1 * 1 KIT
       483,00 €
  • 786-697
    HOOK™-NHS-dPEG4-Biotin Kit
    HOOK™-NHS-dPEG4-Biotin Kit
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 534,00 €
    534,00 €
    1 * 1 KIT
       534,00 €