RNase-DETECT™
Supplier: G-Biosciences
Certificates
About this item
RNase-DETECT™ is a sensitive method to detect RNase contamination.
- Contains aliquots of calibrated RNase substrate for detection of femtogram levels of RNase contamination
- Serial Dilution Ladder (SDL) detection technique supplied for rapid, clear detection of contamination
- Compatible with a wide variety of reagent solutions, including SDS, Tris, salts, agarose
- Suitable to assay solutions that can not be treated with DEPC
Delivery: Supplied with RNase substrate vials, reaction-loading dyes, and protocol for quick analysis of the results.
Frequently Bought Together