Arrest™ extraction buffer
Catalog # 786-133
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:200 Tests
- Method / Format:Precipitation
- Prep size:Scalable
- Refine target molecule:Total RNA
- Sample type:Cultured cellsHuman/animal cells and tissues
- Target molecule:RNA
- Description:Arrest™ Extraction Buffer, for 200×50 mg extractions
- Description:Arrest™ Extraction Buffer
Specifications
About this item
Arrest™ Extraction Buffer is an optimised combination of various chaotropic agents and RNase inhibitors, which inhibits RNase rapidly. It may be used in conjunction with any RNA extraction method, including extractions based on phenol, chloroform and other organic solvents and detergents.
- Rapidly inhibit destructive RNases
- Strong chaotropic buffer for rapid RNA release
- Compatible with numerous species and tissues