Orders LinkContactUsLinkComponent
Capillary tubes
Capillary tubes
Supplier:   HILGENBERG
Capillary tubes
Supplier:  HILGENBERG
Certificates
Capillary tubes
8 Options Available Below
Product Details & Documents
Borosilicate glass, ends cut, with filament.
Product Family Options
Product Information
  • Length
    Ext.Ø
    Ø int.
    Availability
    Price
  • 75 mm
    3 mm
    1,74 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 1 180,00 €
    1 180,00 €
    1 * 100 ST
       1 180,00 €
  • 75 mm
    2 mm
    1 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 929,00 €
    929,00 €
    1 * 100 ST
       929,00 €
  • 75 mm
    2 mm
    1,4 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 553,00 €
    553,00 €
    1 * 100 ST
       553,00 €
  • 75 mm
    1,5 mm
    0,85 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 999,00 €
    999,00 €
    1 * 100 ST
       999,00 €
  • 75 mm
    1,2 mm
    0,69 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 988,00 €
    988,00 €
    1 * 100 ST
       988,00 €
  • 75 mm
    2 mm
    1,6
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 916,00 €
    916,00 €
    1 * 100 ST
       916,00 €
  • 100 mm
    1 mm
    0,75 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 979,00 €
    979,00 €
    1 * 100 ST
       979,00 €
  • 100 mm
    1 mm
    0,58 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 504,00 €
    504,00 €
    1 * 100 ST
       504,00 €