Order Entry
Belgium
ContactUsLinkComponent
Plasmid Screening ToothPick™ and ToothPick™-PCR
Plasmid Screening ToothPick™ and ToothPick™-PCR
Supplier:  G-Biosciences
Certificates
undefined
Plasmid Screening ToothPick™ and ToothPick™-PCR
Supplier:  G-Biosciences

Specifications

  • Method / Format:
    Extraction for direct PCR
  • Prep size:
    Scalable
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    ToothPick™-PCR

Specifications

Related Information

Frequently Bought Together